4-(Cyanomethyl)phenylboronic acid

Supplier: Matrix Scientific

Synonyms: 4-(Cyanomethyl)benzeneboronic acid

027265-1G 027265-5G
102663-654EA 258.46 USD
102663-654 102663-656
4-(Cyanomethyl)phenylboronic acid
4-(Cyanomethyl)phenylboronic acid

Hazard: Irritant

Formula: C₈H₈BNO₂
MW: 160.97 g/mol
MDL Number: MFCD01632200
CAS Number: 91983-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR