3-Chloro-4-morpholino-1,2,5-thiadiazole
Supplier: Matrix Scientific
Synonyms:
Timolol Related Compound F, 4-(4-Chloro-1,2,5-thiadiazol-3-yl)morpholine
027226-1G
027226-5G
102518-800EA
558.46
USD
102518-800
102518-802
3-Chloro-4-morpholino-1,2,5-thiadiazole
3-Chloro-4-morpholino-1,2,5-thiadiazole
Hazard: Irritant
Formula:
C₆H₈ClN₃OS MW: 205.66 g/mol |
MDL Number:
MFCD00274160 CAS Number: 30165-96-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...