1-Methylimidazole-2-carbaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Methyl-1H-imidazole-2-carbaldehyde, 1-Methylimidazole-2-carboxaldehyde
027132-5G
027132-25G
102551-100EA
146.15
USD
102551-100
102551-102
1-Methylimidazole-2-carbaldehyde ≥95%
1-Methylimidazole-2-carbaldehyde
Hazard: Irritant
Formula:
C₅H₆N₂O MW: 110.12 g/mol Boiling Pt: 70-74°C/1mm Melting Pt: 36-39°C Flash Pt: 102°C |
MDL Number:
MFCD01321308 CAS Number: 13750-81-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...