3-Amino-1-adamantanol
Supplier: Matrix Scientific
Synonyms:
3-Aminoadamantan-1-ol
027067-25G
027067-100G
102551-096EA
73.85
USD
102551-096
102551-098
3-Amino-1-adamantanol
3-Amino-1-adamantanol
CAS: 702-82-9
MDL Number: MFCD01821204
Molecular Formula: C10H17NO
Molecular Weight: 167,25
Hazard: Irritant
Formula:
C₁₀H₁₇NO MW: 167.25 g/mol Melting Pt: 263…269 °C (dec.) |
MDL Number:
MFCD01821204 CAS Number: 702-82-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...