4-Chloro-3-iodo-1H-pyrrolo[2,3-b]pyridine ≥95%
Supplier: Matrix Scientific
026242-250MG
026242-1G
102551-092EA
30.77
USD
102551-092
102551-094
4-Chloro-3-iodo-1H-pyrrolo[2,3-b]pyridine ≥95%
4-Chloro-3-iodo-1H-pyrrolo[2,3-b]pyridine
Hazard: Irritant
Formula:
C₇H₄ClIN₂ Melting Pt: 202…204 °C |
MDL Number:
MFCD09266226 CAS Number: 869335-73-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...