trans-4-Methoxycinnamic acid
Supplier: Matrix Scientific
Synonyms:
trans-O-Methyl-p-coumaric Acid, (E)-3-(4-Methoxyphenyl)-2-propenoic acid
025953-100G
025953-500G
102832-930EA
93.85
USD
102832-930
102832-932
trans-4-Methoxycinnamic acid
trans-4-Methoxycinnamic acid
Hazard: Irritant
Formula:
C₁₀H₁₀O₃ MW: 178.19 g/mol |
MDL Number:
MFCD00004398 CAS Number: 943-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...