Monomethyl terephthalate
Supplier: Matrix Scientific
Synonyms:
Methyl hydrogen terephthalate, 4-(Methoxycarbonyl)benzoic acid
024266-25G
024266-100G
102551-054EA
41.54
USD
102551-054
102832-918
Monomethyl terephthalate
Monomethyl terephthalate
Hazard: Irritant
Formula:
C₉H₈O₄ MW: 180.16 g/mol Melting Pt: 220…223 °C |
MDL Number:
MFCD00002557 CAS Number: 1679-64-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...