2-Bromo-p-cresol

Supplier: Matrix Scientific

Synonyms: 2-Bromo-4-methylphenol, 1-Bromo-2-hydroxy-5-methylbenzene

024161-25G 024161-100G
102551-048EA 106.15 USD
102551-048 102551-050
2-Bromo-p-cresol
2-Bromo-p-cresol

Hazard: Irritant

Formula: C₇H₇BrO
MDL Number: MFCD00002151
CAS Number: 6627-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR