2-Bromo-4-chlorophenol
Supplier: Matrix Scientific
024052-25G
024052-100G
102551-044EA
118.46
USD
102551-044
102551-046
2-Bromo-4-chlorophenol
2-Bromo-4-chlorophenol
Hazard: Irritant
Formula:
C₆H₄BrClO |
MDL Number:
MFCD00002319 CAS Number: 695-96-5 UN: 2020 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...