Sulfamethoxazole

Supplier: Matrix Scientific

Synonyms: 4-Amino-N-(5-methylisoxazol-3-yl)benzenesulphonamide

023869-25G 023869-100G
102551-040EA 138.46 USD
102551-040 102551-042
Sulfamethoxazole
Sulfamethoxazole

Hazard: Irritant

Formula: C₁₀H₁₁N₃O₃S
MW: 253.28 g/mol
MDL Number: MFCD00010546
CAS Number: 723-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR