3,4-Dimethylbenzenesulfonyl chloride

Supplier: Matrix Scientific

Synonyms: o-Xylene-4-sulphonyl chloride, 3,4-Dimethylbenzene-1-sulphonyl chloride

021521-25G 021521-100G
102551-030EA 158.46 USD
102551-030 102551-032
3,4-Dimethylbenzenesulfonyl chloride
3,4-Dimethylbenzenesulphonyl chloride

Hazard: Irritant

Formula: C₈H₉ClO₂S
MW: 204.68 g/mol
MDL Number: MFCD00625742
CAS Number: 2905-30-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR