1-Methyl-1H-pyrazol-4-carbaldehyde

Supplier: Matrix Scientific

Synonyms: 1-Methyl-1H-pyrazole-4-carbaldehyde, 1-Methyl-1H-pyrazole-4-carboxaldehyde

021325-1G 021325-5G
102551-014EA 23.08 USD
102551-014 102551-016
1-Methyl-1H-pyrazol-4-carbaldehyde
1-Methyl-1H-pyrazol-4-carbaldehyde

Hazard: Irritant

Formula: C₅H₆N₂O
MW: 110.12 g/mol
MDL Number: MFCD00460465
CAS Number: 25016-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR