1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one ≥98%
Supplier: Matrix Scientific
Synonyms:
7-Deazahypoxanthine
020967-1G
020967-5G
102663-624EA
15.38
USD
102663-624
102663-626
1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one ≥98%
1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one
Hazard: Irritant
Formula:
C₆H₅N₃O MW: 135.13 g/mol Melting Pt: 345…350 °C |
MDL Number:
MFCD08234923 CAS Number: 3680-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...