4-Methyl-2-thenoic acid

Supplier: Matrix Scientific

Synonyms: 4-Methyl-2-thiophenecarboxylic acid, 4-Methylthiophene-2-carboxylic acid

020639-5G 020639-25G
102832-906EA 66.15 USD
102832-906 102832-908
4-Methyl-2-thenoic acid
4-Methyl-2-thenoic acid

Hazard: Irritant

Formula: C₆H₆O₂S
MW: 142.18 g/mol
MDL Number: MFCD00089284
CAS Number: 14282-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR