2-Fluoro-3-formylpyridine ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Fluoronicotinaldehyde
020578-5G
020578-25G
102832-902EA
60
USD
102832-902
102832-904
2-Fluoro-3-formylpyridine ≥97%
2-Fluoro-3-formylpyridine
Hazard: Irritant
Formula:
C₆H₄FNO MW: 125.1 g/mol Boiling Pt: 50 °C (3 mmHg) Density: 1.27 |
MDL Number:
MFCD03095270 CAS Number: 36404-90-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...