4-Chloro-2,6-dimethylpyrimidine
Supplier: Matrix Scientific
018781-1G
018781-5G
102832-874EA
60
USD
102832-874
102832-876
4-Chloro-2,6-dimethylpyrimidine
4-Chloro-2,6-dimethylpyrimidine
Hazard: Irritant
Formula:
C₆H₇ClN₂ MW: 142.03 g/mol |
MDL Number:
MFCD00234197 CAS Number: 4472-45-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...