2,6-Diiodopyridine ≥98%
Supplier: Matrix Scientific
018304-1G
018304-5G
102832-866EA
184.62
USD
102832-866
102832-868
2,6-Diiodopyridine ≥98%
2,6-Diiodopyridine
Hazard: Irritant
Formula:
C₅H₃I₂N MW: 330.89 g/mol Melting Pt: 179…180 °C |
MDL Number:
MFCD07366728 CAS Number: 53710-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...