4-Amino-2-bromophenol ≥98%
Supplier: Matrix Scientific
018180-1G
018180-5G
102663-602EA
69.23
USD
102663-602
102663-604
4-Amino-2-bromophenol ≥98%
4-Amino-2-bromophenol
Hazard: Irritant
Formula:
C₆H₆BrNO |
MDL Number:
MFCD06656565 CAS Number: 16750-67-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...