3-Dimethylamino-2,2-dimethylpropionaldehyde

Supplier: Matrix Scientific

016759-1G 016759-5G
102663-598EA 538.46 USD
102663-598 102663-600
3-Dimethylamino-2,2-dimethylpropionaldehyde
3-Dimethylamino-2,2-dimethylpropionaldehyde

Hazard: Irritant

Formula: C₇H₁₅NO
MW: 129.2 g/mol
MDL Number: MFCD00043600
CAS Number: 15451-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR