7-Bromo-2-naphthol ≥98%
Supplier: Matrix Scientific
016591-1G
016591-5G
102663-594EA
60
USD
102663-594
102663-596
7-Bromo-2-naphthol ≥98%
7-Bromo-2-naphthol
Hazard: Irritant
Formula:
C₁₀H₇BrO |
MDL Number:
MFCD00277644 CAS Number: 116230-30-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...