2-Chloro-4-pyridineboronic acid pinacol ester
Supplier: Matrix Scientific
Synonyms:
2-Chloropyridine-4-boronic acid pinacol ester, 2-(2-Chloro-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Chloropyridine-4-boronic acid Pinacol ester
016056-1G
016056-5G
102550-980EA
203.08
USD
102550-980
102550-982
2-Chloro-4-pyridineboronic acid pinacol ester
2-Chloro-4-pyridineboronic acid pinacol ester
Hazard: Irritant/Store in Freezer
Formula:
C₁₁H₁₅BClNO₂ MW: 239.51 g/mol Melting Pt: 64.7-64.8°C |
MDL Number:
MFCD04039870 CAS Number: 458532-84-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...