4-Methoxypiperidine hydrochloride

Supplier: Matrix Scientific

015567-1G 015567-5G
102832-854EA 44.62 USD
102832-854 102832-856
4-Methoxypiperidine hydrochloride
4-Methoxypiperidine hydrochloride

Hazard: Irritant

Formula: C₆H₁₄ClNO
Melting Pt: 137…139 °C
MDL Number: MFCD06800959
CAS Number: 4045-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR