Isopropyl-4-aminobenzoate
Supplier: Matrix Scientific
Synonyms:
Isopropyl 4-aminobenzoate
011329-1G
011329-5G
102550-962EA
52.31
USD
102550-962
102550-964
Isopropyl-4-aminobenzoate
Isopropyl-4-aminobenzoate
Hazard: Irritant
Formula:
C₁₀H₁₃NO₂ MW: 179.21 g/mol Boiling Pt: 190-192°C/0.5 Torr Melting Pt: 86°C |
MDL Number:
MFCD00017108 CAS Number: 18144-43-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...