4-(4-Ethylpiperazin-1-yl)phenylamine
Supplier: Matrix Scientific
Synonyms:
4-(4-Ethylpiperazin-1-yl)aniline
010871-1G
010871-5G
102550-954EA
158.46
USD
102550-954
102550-956
4-(4-Ethylpiperazin-1-yl)phenylamine
4-(4-Ethylpiperazin-1-yl)phenylamine
Hazard: Irritant
Formula:
C₁₂H₁₉N₃ MW: 205.3000 g/mol Melting Pt: 73…75 °C |
MDL Number:
MFCD00702246 CAS Number: 115619-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...