1H-Tetrazole-5-acetic acid ≥95%
Supplier: Matrix Scientific
010607-1G
010607-5G
102832-822EA
76.92
USD
102832-822
102832-824
1H-Tetrazole-5-acetic acid ≥95%
1H-Tetrazole-5-acetic acid
Hazard: Irritant
Formula:
C₃H₄N₄O₂ MW: 128.09 g/mol Melting Pt: 127 °C |
MDL Number:
MFCD03094010 CAS Number: 21743-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...