2-(Piperidin-4-yl)-1H-benzo[d]imidazole
Supplier: Matrix Scientific
010422-1G
010422-5G
102832-818EA
93.85
USD
102832-818
102832-820
2-(Piperidin-4-yl)-1H-benzo[d]imidazole
2-(Piperidin-4-yl)-1H-benzo[d]imidazole
Hazard: Irritant
Formula:
C₁₂H₁₅N₃ Melting Pt: 230 °C (dec.) |
MDL Number:
MFCD01475843 CAS Number: 38385-95-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...