2-Amino-5-bromobenzamide ≥97%

Supplier: Matrix Scientific

009823-1G 009823-5G
102550-944EA 53.85 USD
102550-944 102550-946
2-Amino-5-bromobenzamide ≥97%
2-Amino-5-bromobenzamide

Hazard: Irritant

Formula: C₇H₇BrN₂O
Melting Pt: 189…191 °C
MDL Number: MFCD04037881
CAS Number: 16313-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR