3,5-Dimethylisoxazole-4-carboxylic acid
Supplier: Matrix Scientific
Synonyms:
3,5-Dimethylisoxasole-4-carboxylic acid
008380-5G
008380-25G
102550-916EA
106.15
USD
102550-916
102550-918
3,5-Dimethylisoxazole-4-carboxylic acid
3,5-Dimethylisoxazole-4-carboxylic acid
Hazard: Irritant
Formula:
C₆H₇NO₃ MW: 141.13 g/mol Melting Pt: 42-43°C |
MDL Number:
MFCD00051657 CAS Number: 2510-36-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...