4-Amino-3-phenylbutyric acid hydrochloride
Supplier: Matrix Scientific
Synonyms:
4-Amino-3-phenylbutanoic acid
008053-1G
008053-5G
102550-912EA
110.77
USD
102550-912
102550-914
4-Amino-3-phenylbutyric acid hydrochloride
4-Amino-3-phenylbutyric acid hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₄ClNO₂ MW: 215.68 g/mol Melting Pt: 253 °C (dec.) |
MDL Number:
MFCD00075860 CAS Number: 1078-21-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...