5-Methoxy-3-pyridineboronic acid pinacol ester ≥98%
Supplier: Matrix Scientific
Synonyms:
5-Methoxy-3-pyridylboronic acid Pinacol ester, 3-Methoxypyridine-5-boronic acid Pinacol ester, 3-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(5-Methoxy-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
007384-1G
007384-5G
102550-902EA
126.15
USD
102550-902
102550-904
5-Methoxy-3-pyridineboronic acid pinacol ester ≥98%
5-Methoxy-3-pyridineboronic acid pinacol ester
Hazard: Irritant/Store in Freezer
Formula:
C₁₂H₁₈BNO₃ |
MDL Number:
MFCD03412068 CAS Number: 445264-60-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...