3-Amino-2-chloro-6-(trifluoromethyl)pyridine ≥98%

Supplier: Matrix Scientific

006910-1G 006910-5G
102832-782EA 63.08 USD
102832-782 102832-784
3-Amino-2-chloro-6-(trifluoromethyl)pyridine ≥98%
3-Amino-2-chloro-6-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C6H4ClF3N2
Melting Pt: 92…94 °C
MDL Number: MFCD01862661
CAS Number: 117519-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR