4-(Tetrahydropyran-2-yloxy)phenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(2-Tetrahydropyranyloxy)benzeneboronic acid, 4-(Tetrahydro-2H-pyran-2-yloxy)phenylboronic acid, 4-(Tetrahydro-2H-pyran-2-yloxy)benzeneboronic acid, (4-((Tetrahydro-2H-pyran-2-yl)oxy)phenyl)boronic acid

006780-5G 006780-25G
102550-884EA 36.92 USD
102550-884 102550-886
4-(Tetrahydropyran-2-yloxy)phenylboronic acid ≥95%
4-(Tetrahydropyran-2-yloxy)phenylboronic acid

Hazard: Irritant/Refrigerate

Formula: C₁₁H₁₅BO₄
MW: 222.05 g/mol
MDL Number: MFCD02183525
CAS Number: 182281-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR