Bis(neopentyl glycolato)diboron ≥97%
Supplier: Matrix Scientific
Synonyms:
5,5,5',5'-Tetramethyl-2,2'-bi(1,3,2-dioxaborinane), 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane
006754-25G
006754-100G
102550-880EA
52.31
USD
102550-880
102550-882
Bis(neopentyl glycolato)diboron ≥97%
Bis(neopentyl glycolato)diboron
Hazard: Irritant/Refrigerate
Formula:
C₁₀H₂₀B₂O₄ Melting Pt: 170…78 °C |
MDL Number:
MFCD02093062 CAS Number: 201733-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...