3-Chloro-2-fluorobenzyl alcohol

Supplier: Matrix Scientific

002096-5G 002096-25G
102550-810EA 181.54 USD
102550-810 102550-812
3-Chloro-2-fluorobenzyl alcohol
3-Chloro-2-fluorobenzyl alcohol

Hazard: Irritant

Formula: C₇H₆ClFO
MW: 160.58 g/mol
Melting Pt: 39…41 °C
MDL Number: MFCD01631549
CAS Number: 261723-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR