Viable Pea Seeds
Supplier: CROSMAN SEED CORPORATION
820LB 821*1 OZ.
470007-182EA 8.5 USD
470007-182 470105-818
Viable Pea Seeds
Specimens Live Specimens Plants - Living Material
Learn genetics like Mendel.
  • Seeds of multiple Phenotypes
  • Variable sizes for each seed type
Use these peas in any lab activity, not just genetics. Peas are an excellent option for germination, growth, and genetic experiments.
Información accesorios: Where a Count of Seeds is indicted, the Quantity is an estimate based on Weight and may vary slightly.