Cancer Stem Cell Medium
Supplier: Promocell, Inc.
C-28070 C-28075
75794-940EA 292.04 USD
75794-940 76321-026
Cancer Stem Cell Medium
Cell Culture Media
PromoCell 3D Tumorsphere Medium has been designed to meet your requirements for the extended serial 3D tumorsphere culture. 3D Tumorsphere Medium supports the tumorsphere/mammosphere culture of the most commonly used cancer cell lines.
  • Directly switch from 2D to 3D culture
  • Supports most commonly used cancer cell lines
  • Allows extended serial tumorsphere passages (>10)
  • High tumorsphere formation efficiency
  • Serum-free and chemically defined
  • Ready to use
The PromoCell cancer stem cell medium is a culture system designed for culture of sphere-forming cancer cell lines as tumorspheres/mammospheres. The formulation supports sustained cell proliferation, allowing for serial passage of the 3D culture.
The PromoCell cancer stem cell medium is ready-to-use and chemically defined providing a culture environment devoid of all stimuli originating from non-defined materials. It is particularly suitable as a cost-efficient tool for the standardized routine-culture of tumorspheres/mammospheres.
List of cell types tested for serial passage with the PromoCell: Bran U-87 MG, Breast MCF-7; Colon HT-29, Connective tissue HT1080; Liver HepG2; Lung A-549; Pancreas Panc-1; Prostate LNCaP; Skin A-431.
Información para pedidos: PromoCell cancer stem cell medium is available as medium (Ready to- use) and consists of a bottle of Basal medium and one vial of Supplement- Mix. adding the SupplementMix to the Basal medium results in the complete medium.