TSK-Gel® Universal Reverse Phase Columns, Tosoh Biosciences
Supplier: Tosoh Bioscience
21458 21459 21460 21462 21995 22702 22705 22707 22716 22719 22724 22728 22729 22735 22740 22747 22748 22752 22755 22760 22762 21831 21940 22725 22763 21812 22712 22721 22741 21938 22703 22738 22700 22739 22701 22706 22710 22711 22723 22731 22717 22754 21457 21810 21814 21830 21838 21997 22708 22709 22718 22720 22727 22730 22733 22744 22750 22751 22757 22761 22764 22765 21453 21455 21456 21461 21811 21813 21829 21839 21841 21996 22704 22713 22714 22715 22722 22726 22732 22734 22736 22737 22742 22743 22745 22746 22749 22753 22756 22758 22759
101742-624EA 1154.05 USD
101742-624 101742-626 101742-628 101742-632 101742-792 101742-804 101742-810 101742-814 101742-832 101742-838 101742-848 101742-856 101742-858 101742-870 101742-880 101742-894 101742-896 101742-904 101742-910 101742-920 101742-924 101742-676 101742-744 101742-850 101742-926 101742-662 101742-824 101742-842 101742-882 101742-742 101742-806 101742-876 101742-800 101742-878 101742-802 101742-812 101742-820 101742-822 101742-846 101742-862 101742-834 101742-908 101742-622 101742-658 101742-666 101742-674 101742-682 101742-796 101742-816 101742-818 101742-836 101742-840 101742-854 101742-860 101742-866 101742-888 101742-900 101742-902 101742-914 101742-922 101742-928 101742-930 101742-614 101742-618 101742-620 101742-630 101742-660 101742-664 101742-672 101742-684 101742-686 101742-794 101742-808 101742-826 101742-828 101742-830 101742-844 101742-852 101742-864 101742-868 101742-872 101742-874 101742-884 101742-886 101742-890 101742-892 101742-898 101742-906 101742-912 101742-916 101742-918
TSK-Gel® Universal Reverse Phase Columns, Tosoh Biosciences
Chromatography Columns
TSK-Gel® ODS-100 Universal reverse phase columns incorporate good surface properties to limit secondary interactions of basic, acidic and chelating compounds.
  • They offer high efficiency and symmetrical peak shape
  • The columns are suitable for demanding separations in quality control as well as in R and D
  • General purpose RP columns have ultra high purity Type B base silica combined with monomeric bonding
TSK-Gel® ODS-100V columns are general purpose columns providing strong retention and high selectivity for polar solutes
TSK-Gel® ODS-100Z RP columns contain a high density monomeric C18 bonded phase for maximum retention and selectivity of small molecular weight compounds. Exhaustive endcapping prevents secondary interaction with residual silanol groups.
TSK-Gel® ODS-100 columns are available in 3 and 5 µm particle size. 3 µm columns, providing fast and highly efficient separation, are well suited for LC/MS applications.
Unique, highly efficient bonding and endcapping procedures. Monomeric bonded phase chemistry provides complete wetting and retention stability in 100% aqueous mobile phase.