SafeCup™ lll MultiDrug Panel, Germaine® Laboratories
Supplier: Germaine Laboratories
40675 40105 45485 45475 41577
75796-182CS 298.74 USD
75796-182 75796-216 75796-218 75796-236 75796-238 75799-284
SafeCup™ lll MultiDrug Panel, Germaine® Laboratories
Drug Tests
SafeCup™ lll Round Cups are a series of All-in-One (collection/testing/transport) qualitative tests for the detection of drugs of abuse in urine
Tests include 25 collection cups, security seals, gloves, and instructions.
Tests adhere to SAMSHA cutoff levels. Cups feature a leak-proof cap, temperature strip, and security seal included.
Certificaciones: CLIA Waived.