Toyopearl® Hydrophobic Interaction Chromatography Resins, Tosoh Bioscience
Supplier: TOSOH BIOSCIENCE, LLC MS
43125 43278 43150 43175 19806
100367-312EA 1015.3 USD
100367-312 100368-004 100368-168 100367-418 100367-532
Toyopearl® Hydrophobic Interaction Chromatography Resins, Tosoh Bioscience
Chromatography Sorbents HPLC Sorbents
TOYOPEARL and TSKgel LabPak media products are small package sizes of TOYOPEARL and TSKgel bulk media products
The larger resin amounts in LabPak products allow the packing of wider bore and longer columns. This helps to accurately determine the resin's dynamic binding capacity, selectivity, column efficiency and operating conditions.
They contain three or four different ligand types offered for a particular chromatography mode.