Guanidinium carbonate, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.20611.9025
8.20611.9050
EM8.20611.9025EA
1189
USD
EM8.20611.9025
EM8.20611.9050
Guanidinium carbonate, Sigma-Aldrich®
Guanidinium carbonate
Formula:
C₃H₁₂N₆O₃ Melting Pt: 231 °C Density: 1.251 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013029 CAS Number: 593-85-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...