Myeloperoxidase, MPO, Human ELISA, BioVendor
Supplier: BioVendor
RHK324-02R RHK324-01R
102552-488EA 1384.62 USD
102552-488 101189-724
Myeloperoxidase, MPO, Human ELISA, BioVendor
Assays ELISAs
MPO is an important enzyme used during phagocytic lysis of engulfed foreign particles.
  • Calibration Range: 0.4-100 ng/mL
  • Limit of Detection: 0.4 ng/mL
  • Plasma, Sputum: 100 uL/well
Myeloperoxidase (MPO) is a glycoprotein with a alpha2beta2 heteromultimer expressed in all cells of the myeloid linage. MPO is abundantly present in azurophilic granules of polymorphonuclear neutrophils. It is an important enzyme used during phagocytic lysis of engulfed foreign particles which takes part in the defense of the organism through production of hypochlorous acid (HOCl), a potent oxidant. MPO is rapidly released by activated polymorphonuclear neutrophils. Involvement of MPO has been described in numerous diseases such as atherosclerosis, lung cancer, Alzheimer's disease and multiple sclerosis. Autoimmune antibodies to MPO are involved in Wegener’s disease. Since the discovery of MPO deficiency, initially regarded as rare and restricted to patients suffering from severe infections, MPO has attracted more clinical attention.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.