Sensiscript RT Kit , Qiagen
205211 205213
220006-680EA 638.46 USD
220006-680 220006-604
Sensiscript RT Kit , Qiagen
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Sensiscript RT kit for reverse transcription of less than 50ng RNA per reaction for end-point P.
  • Sensitive detection of as few as 10 copies of templates
  • High cDNA yields due to high-affinity enzyme
  • No additional RNase H digestion step required
  • Fast and easy procedure with no pipetting steps
Includes Sensiscript Reverse Transcriptase, 150L 10x Buffer RT, 100L dNTP Mix (contains 5mM each of dNTP), and 1.1mL RNase-free water.