RNAlater RNA Stabilization Reagent , Qiagen
76106 76104
220002-448EA 767.69 USD
220002-448 220002-450
RNAlater RNA Stabilization Reagent , Qiagen
Protein Expression Protein Expression Reagents
RNAlater RNA Stabilization Reagent for immediate stabilization of RNA in tissues.
  • Immediate RNA stabilization and protection
  • Reliable gene expression and gene-profiling data
  • Convenient and safe handling at room temperature
  • No need for liquid nitrogen or dry ice
  • Tissue archiving without risk of RNA degradation
Ideal for total RNA, miRNA, and mRNA purification.