REPLI-g Mini Kit , Qiagen
150023 150025
220006-812EA 484.62 USD
220006-812 220006-748
REPLI-g Mini Kit , Qiagen
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
REPLI-g Mini kit for highly uniform whole-genome amplification from small or precious samples.
  • Easy amplification with consistent yields of 10µg
  • Unbiased amplification of genomic loci
  • Reliable results due to Multiple Displacement Amplification (MDA)
  • Amplified DNA highly suitable for most downstream applications
  • No risk of DNA degradation during long-term storage
Includes DNA Polymerase, Buffers and Reagents for 25 x 50L whole-genome amplification reactions.