QuantiTect Virus Kit, Qiagen
211015 211011 211013
220006-030EA 7520 USD
220006-030 102090-580 102090-582
QuantiTect Virus Kit, Qiagen
Assays qPCR Assays
QuantiTect Virus kits for highly sensitive detection of viral RNA and/or DNA.
  • High sensitivity in single and multiplex assays
  • Detection of viral RNA and/or DNA in the same reaction
  • Clear detection of weak positive signals
  • Fast, universal two-step protocols
  • 5x master mix for higher sensitivity with more sample input
Includes QuantiTect Virus Master Mix (contains ROX dye), QuantiTect Virus RT Mix, RNase-free water, and QuantiTect Nucleic Acid Dilution Buffer.