Omniscript RT Kit , Qiagen
205113 205111
220006-678EA 2076.92 USD
220006-678 220006-758
Omniscript RT Kit , Qiagen
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Omniscript RT kit for reverse transcription of 50ng to 2μg RNA per reaction for end-point PCR.
  • High cDNA yields due to high-affinity enzyme
  • Fast and easy procedure
  • No pipetting steps
  • Sensitive detection of as few as 10 copies of template
  • No additional RNase H digestion step required
Kit contents: 40U Omniscript Reverse Transcriptase, 30L 10x buffer RT, 20L dNTP mix (contains 5mM each dNTP), 1.1mL RNase-free water.