Taq PCR Core Kit, Qiagen
201225 201223
220006-546EA 1066.15 USD
220006-546 220006-602
Taq PCR Core Kit, Qiagen
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Taq PCR Core Kit for standard and specialized applications, with included dNTP solution set.
  • Ideal for standard and specialized PCR applications
  • Ready-to-load PCR buffer for faster handling
  • Convenient choice of formats
  • Easy to handle
Includes Taq DNA Polymerase, 10x PCR Buffer, 10x CoralLoad PCR Buffer, 5x Q-Solution, 25mM MgCl2, dNTP Mix. QIAGEN PCR Buffer for minimal optimization. Additional ready-to-load PCR buffer for faster handling. Q-Solution for amplification of GC-rich templates. Choice of formats for convenience and ease of handling.