Forma™ Steri-Cult™ CO₂ Incubators, Thermo Scientific
Supplier: Thermo Fisher Scientific
3310 3308 3307
101623-114EA 37538.46 USD
101623-114 101623-140 101623-138
Forma™ Steri-Cult™ CO₂ Incubators, Thermo Scientific
Incubators CO₂ Incubators
These stackable Forma™ Steri-Cult™ CO2 Incubators allow user to carry out high-volume culturing with a high level of contamination control.
  • Optimized airflow system will not interfere with samples or incubator function
  • In-chamber HEPA air filtration continuously removes particulates for Class 100 quality conditions
  • Audible alarm activates if outer door is opened during the cycle or if temperature reaches 60°C or greater
  • Alphanumeric messaging center clearly displays temperature, CO2 and RH
  • Access code prevents accidental initiation of the cycle
This incubator combines the most advanced components available into a single incubator: CO2 sensor, precise microprocessor controls, active humidity control, HEPA air filtration and high-temperature decontamination capability.