Nut, Swagelok®, Restek
Supplier: Restek
23152 23100 23101 23102 23150 23151
10057-918PK 69.8 USD
10057-918 10058-866 10058-868 10058-870 10058-922 10058-924
Nut, Swagelok®, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
No coating required, but nuts can be treated upon request (custom parts).
Información para pedidos: Note: Nuts and ferrules are not treated unless requested (custom parts). Nuts and ferrules normally are not in contact with sample pathway, and thus do not require coating.