Frit Inlet Liner with Top Taper for Bruker/Varian GCs Equipped with 1078/1079 inlets, Restek
Supplier: Restek
21709 21709-214
RK21709PK 260.49 USD
RK21709 75778-636
Frit Inlet Liner with Top Taper for Bruker/Varian GCs Equipped with 1078/1079 inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
Superior coverage is delivered even with highly active compounds.